For-Met-Leu-Phe-OH
Supplier: BACHEM AMERICAS INC
4001509.0250 4001509.1000
H-3030.0250BAEA 429.44 USD
H-3030.0250BA H-3030.1000BA
For-Met-Leu-Phe-OH
Proteins and Peptides
There is evidence that pretreatment of human polymorphonuclear neutrophils (PMN) with H-3030 results in the inhibition of TNF-α secretion, which indicates that this typical proinflammatory agent could play, at least at determined conditions, an antiinflammatory role. See also the Nle analog (H-3060).