beta-Amyloid (40-1) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4030203.0500
4030203.1000
H-2972.0500BAEA
401.67
USD
H-2972.0500BA
H-2972.1000BA
beta-Amyloid (40-1) Trifluoroacetate
Proteins and Peptides
Reverse sequence of Aβ 1-40. Inactive control for H-1194 and H-5568.