Chicken;Human;Mouse;Pig;Rat;Sheep PACAP-38 (6-38) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4031158.0500 4031158.1000
H-2734.0500BAEA 408.5 USD
H-2734.0500BA H-2734.1000BA
Chicken;Human;Mouse;Pig;Rat;Sheep PACAP-38 (6-38) Trifluoroacetate
Proteins and Peptides
The PACAP selective antagonist PACAP (6-38) is a potent and selective inhibitor of PACAP-27-stimulated pituitary adenylate cyclase with a Ki values of 150 nM.