BQ-788
Supplier: BACHEM AMERICAS INC
4026108.0001 4026108.0005
H-2492.0001BAEA 435.71 USD
H-2492.0001BA H-2492.0005BA
BQ-788
Proteins and Peptides
Endothelins are peptides with exceptional vasoconstrictor potency. They play an important role in intercellular communications.
In vitro, BQ-788 potently and competitively inhibited ¹²⁵I-labeled ET-1 binding to ET-B receptors on human Girardi heart cells (IC₅₀= 1.2 nM). However, BQ-788 only poorly inhibited ET-1 binding to ET-A receptors on human neuroblastoma cell line SK-N-MC cells (IC₅₀= 1300 nM). Thus the ET-B receptor antagonist BQ788 may be considered as a powerful tool for investigating the role of ET in physiological and pathological processes.