Gluten Exorphin B5
Supplier: BACHEM AMERICAS INC
4025007.0025
4025007.0100
H-1666.0025BAEA
735.3
USD
H-1666.0025BA
H-1666.0100BA
Gluten Exorphin B5
Proteins and Peptides
Besides deltorphins, dermorphins, dynorphins, enkephalins and endorphins, Bachem offers opioid peptides such as acetalins, BAM peptides, α-casein and gluten exorphins, endomorphins, kyotorphins, metorphamide, opiorphins, δ opioid receptor antagonists, and valorphin.
Among all exogenous opioid peptides reported to date and especially among the gluten exorphins A, B and C, gluten exorphin B5 shows the most potent activity. Its IC₅₀ values were 0.05 µM and 0.017 µM in the GPI and MVD assays, respectively.