H-Ile-Pro-OH
Supplier: BACHEM AMERICAS INC
4002494.0001
4002494.0005
G-2425.0001BAEA
441.71
USD
G-2425.0001BA
G-2425.0005BA
H-Ile-Pro-OH
Proteins and Peptides
See also H-3974, H-Lys(4-nitro-Z)-pyrrolidide · HCl (N-1710) and H-Phe-pyrrolidide (E-3275).
Competitive inhibitor of porcine dipeptidyl peptidase IV (DPP IV), IC₅₀ 0.015 mM.