H-His-Gly-OH
Supplier: BACHEM AMERICAS INC
4001227.0001 4001227.0005
G-2305.0001BAEA 653.59 USD
G-2305.0001BA G-2305.0005BA
H-His-Gly-OH
Proteins and Peptides
Short histidine-containing peptides can form stable complexes with copper and other transition metals. See also His-Leu (G-2310) and the product family 'Liver Cell Growth Factor (GHK) and Analogs'.
The dipeptide L-histidylglycine forms ESR-active Cu(II) complexes.