H-Glu-Gly-OH

Supplier: BACHEM AMERICAS INC

4004492.0001 4004492.0005
G-1935.0001BAEA 441.58 USD
G-1935.0001BA G-1935.0005BA
H-Glu-Gly-OH
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/Peptide Name H-Glu-Gly-OH
CAS No. 13716-89-7
MDL No. MFCD00037311
Sequence H-Glu-Gly-OH
Molecular Weight 204.18
Molecular Formula C₇H₁₂N₂O₅
Storage Conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR