(H-Cys-Gly-OH)₂
Supplier: BACHEM AMERICAS INC
4002368.0250
4002368.1000
G-1815.0250BAEA
361.17
USD
G-1815.0250BA
G-1815.1000BA
(H-Cys-Gly-OH)₂
Proteins and Peptides
Please see also 5-(Pentafluorobenzoylamino)fluorescein (M-2375), a sensitive fluorogenic substrate for the determination of glutathione S-transferase (GST) activity and glutathione (GSH) concentration, and the carba-analog of GSH, ophthalmic acid, H-3145.
NSC333711. Besides its reduced form, this product of glutathione metabolism is found in plasma.