LB Agar Plate with Ampicillin, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J61314-EQF J63197-EQF J63497-EQF
AAJ61314-EQFEA 313.76 USD
AAJ61314-EQF AAJ63197-EQF AAJ63497-EQF
LB Agar Plate with Ampicillin, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology

This LB Agar plate with ampicillin is used with recombinant strains of E.coli containing plasmids with the resistance gene to Ampicillin such as pBluescript and pUC series of plasmids.


  • Solid
  • Soluble in water


Storage Temperature: 2°C to 8°C Physical Form: Solid


Precaución: Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR