LB Agar Plate with Chloramphenicol, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J61140-EQF J62923-EQF
AAJ61140-EQFEA 213 USD
AAJ61140-EQF AAJ62923-EQF
LB Agar Plate with Chloramphenicol, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology
This Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E.coli.
It is used with recombinant strains of E.coli containing plasmids with the resistance gene to 25 µg/mL chloramphenicol such as pBluescript, pGEM, or the pUC series of plasmids.
Precaución: Keep away from strong oxidizing agents and in a container tightly closed in a dry and well-ventilated place.