2-(Aminomethyl)piperidine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Piperidylmethylamine
L20122-06
L20122-14
AAL20122-06EA
44.7
USD
AAL20122-06
AAL20122-14
2-(Aminomethyl)piperidine 98%
2-(Aminomethyl)piperidine
Purity: 98%
CAS No.: 22990-77-8
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 68°(154°F)
Refractive Index: 1.4850
Boiling Point: 63-66°/12mm
Beilstein Reference: 102994
UN No.: 2735
MDL No.: MFCD00129011
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 63…66 °C/12 mm Density: 0.942 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00129011 CAS Number: 22990-77-8 EINECS: 245-371-1 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...