trans,trans-Farnesol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2E,6E)-3,7,11-Trimethyldodeca-2,6,10-trien-1-ol
L14348-03
L14348-06
L14348-14
AAL14348-03EA
25.5
USD
AAL14348-03
AAL14348-06
AAL14348-14
trans,trans-Farnesol 97%
trans,trans-Farnesol
Purity: 97%
CAS No.: 106-28-5
Molecular Formula: C15H26O
Formula Weight: 222.37
Flash Point: 96°(204°F)
Refractive Index: 1.4900
Boiling Point: 137°/3mm
Beilstein Reference: 1723039
Merck Reference: 14,3937
MDL No.: MFCD00002918
Formula:
C₁₅H₂₆O MW: 222.37 g/mol Boiling Pt: 137 °C/3 mm Density: 0.886 Flash Pt: 96 °C(204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002918 CAS Number: 106-28-5 EINECS: 225-004-1 Merck Index: 14,3937 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...