7,8-Dihydroxy-4-methylcoumarin 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methyldaphnetin, 7,8-Dihydroxy-4-methyl-2H-chromen-2-one
L14069-03
L14069-ME
AAL14069-03EA
52.4
USD
AAL14069-03
AAL14069-MC
AAL14069-ME
7,8-Dihydroxy-4-methylcoumarin 97%
7,8-Dihydroxy-4-methylcoumarin
Can be used as a fluorescent sensor to monitor the consumption of a boronic acid in a Suzuki cross-coupling reaction. Use a long wave UV lamp.
Purity: 97%
CAS No.: 2107-77-9
Molecular Formula: C10H8O4
Formula Weight: 192.17
Storage Temperature: 2°C to 8°C
Melting Point: 241-244°
Beilstein Reference: 177613
MDL No.: MFCD00016972
Formula:
C₁₀H₈O₄ MW: 192.17 g/mol Melting Pt: 241…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016972 CAS Number: 2107-77-9 EINECS: 218-290-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...