Cinnamyl anthranilate 99%
Supplier: Thermo Scientific Chemicals
L13659-06
L13659-14
AAL13659-06EA
59.7
USD
AAL13659-06
AAL13659-14
Cinnamyl anthranilate 99%
Cinnamyl anthranilate
Purity: 99%
CAS No.: 87-29-6
Molecular Formula: C16H15NO2
Formula Weight: 253.30
Melting Point: 64-66°
Boiling Point: 322°
Beilstein Reference: 2733435
MDL No.: MFCD00053631
Formula:
C₁₆H₁₅NO₂ MW: 253.30 g/mol Boiling Pt: 322 °C Melting Pt: 64…66 °C Density: 1.180 Storage Temperature: Ambient |
MDL Number:
MFCD00053631 CAS Number: 87-29-6 EINECS: 201-738-8 Merck Index: 14,2303 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...