5-(2-Thienyl)pentanoic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-(Thiophen-2-yl)pentanoic acid
L13122-03
L13122-06
AAL13122-03EA
50.2
USD
AAL13122-03
AAL13122-06
5-(2-Thienyl)pentanoic acid 95%
5-(2-Thienyl)pentanoic acid
Purity: 95%
CAS No.: 21010-06-0
Molecular Formula: C9H12O2S
Formula Weight: 184.26
Melting Point: 39-42°
Beilstein Reference: 126021
MDL No.: MFCD00151849
Formula:
C₉H₁₂O₂S MW: 184.26 g/mol Melting Pt: 39…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151849 CAS Number: 21010-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...