2-(Trimethylsilylethynyl)pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Pyridylethynyl)trimethylsilane, 2-[(Trimethylsilyl)ethynyl]pyridine, 2-((Trimethylsilyl)ethynyl)pyridine
L11251-03
L11251-06
AAL11251-03EA
48.1
USD
AAL11251-03
AAL11251-06
2-(Trimethylsilylethynyl)pyridine 97%
2-(Trimethylsilylethynyl)pyridine
Purity: 97%
CAS No.: 86521-05-3
Molecular Formula: C10H13NSi
Formula Weight: 175.31
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5330
Melting Point: 40-43°
Boiling Point: 85-86°/1mm
Beilstein Reference: 4176916
MDL No.: MFCD00066349
Formula:
C₁₀H₁₃NSi Boiling Pt: 85…86 °C/1 mm Melting Pt: 40…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00066349 CAS Number: 86521-05-3 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...