Hydriodic acid 57% (w/w) in aqueous solution stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: stab. with hypophosphorous acid 1.5%

L10410-22 L10410-36
AAL10410-22EA 83.92 USD
AAL10410-22 AAL10410-36
Hydriodic acid 57% (w/w) in aqueous solution stabilized
Hydriodic acid

CAS No.: 10034-85-2 Molecular Formula: HI Formula Weight: 127.91 Physical Form: Liquid Density: 1.701 Sensitivity: Light Sensitive Appearance: Colorless to yellow Odor: Pungent Boiling Point: 126-127° Merck Reference: 14,4776 UN No.: 1787 MDL No.: MFCD00011347

Formula: HI
Boiling Pt: 126…127 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011347
CAS Number: 10034-85-2
EINECS: 233-109-9
UN: 1787
ADR: 8,III
Merck Index: 14,4776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR