Dihydrofluorescein diacetate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-(3,6-Diacetoxy-9H-xanthen-9-yl)benzoic acid
J65759-03
J65759-MD
AAJ65759-03EA
222
USD
AAJ65759-03
AAJ65759-MD
Dihydrofluorescein diacetate 97%
Dihydrofluorescein diacetate
Fluorescent dye for detecting intracellular oxidants
Purity: 97%
CAS No.: 35340-49-9
Molecular Formula: C24H18O7
Formula Weight: 418.39
Physical Form: Powder
Appearance: White to yellow
Melting Point: 213-215°
Beilstein Reference: 364167
MDL No.: MFCD00005056
Applications: Fluorescent dye for detecting intracellular oxidants
Formula:
C₂₄H₁₈O₇ MW: 418.401 g/mol Melting Pt: 213…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005056 CAS Number: 35340-49-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...