2'-Amino-2'-deoxyadenosine 98%
Supplier: Thermo Scientific Chemicals
J65532-03
J65532-MD
AAJ65532-03EA
492
USD
AAJ65532-03
10815-326
2'-Amino-2'-deoxyadenosine 98%
2'-Amino-2'-deoxyadenosine
Powder
Purity: 98%
CAS No.: 10414-81-0
Molecular Formula: C10H14N6O3
Formula Weight: 266.26
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD06657636
Formula:
C₁₀H₁₄N₆O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD06657636 CAS Number: 10414-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...