2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
Supplier: Thermo Scientific Chemicals
J64981-LB0
J64981-MA
AAJ64981-LB0EA
76.7
USD
AAJ64981-LB0
AAJ64981-MA
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
2-(3-(2,3-Dichlorophenoxy)propylamino)ethanol hydrochloride
Powder
CAS No.: 418788-90-6
Molecular Formula: C11H15Cl2NO2.HCl
Formula Weight: 300.61
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at ~25 mg/ml. Also soluble in ethanol, DMSO or DMF at ~20mg/ml. Soluble in PBS, pH 7.2 at ~10 mg/ml.
MDL No.: MFCD06410993
Formula:
C₁₁H₁₅Cl₂NO₂·HCl MW: 300.61 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06410993 CAS Number: 418788-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...