uPA Inhibitor II, solid

Supplier: Thermo Scientific Chemicals

J64874-LB0 J64874-MB
AAJ64874-LB0EA 206 USD
AAJ64874-LB0 AAJ64874-MB
uPA Inhibitor II, solid
uPA Inhibitor II

A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator


  • Light Sensitive


Yellow solid


A cell-permeable oxazolidinone-carboxamidine compound that acts as a potent, selective, and active-site targeting inhibitor of urokinase-type plasminogen activator

Formula: C₁₇H₁₃N₃O₂·C₂HF₃O₂
MW: 405.3 g/mol
Storage Temperature: Ambient
CAS Number: 1186653-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR