2'-Amino-2'-deoxyguanosine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64650-03
J64650-MD
AAJ64650-03EA
619
USD
AAJ64650-03
10815-310
2'-Amino-2'-deoxyguanosine 98%
2'-Amino-2'-deoxyguanosine
Powder
Purity: 98%
CAS No.: 60966-26-9
Molecular Formula: C10H14N6O4
Formula Weight: 282.26
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD01723954
Formula:
C₁₀H₁₄N₆O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD01723954 CAS Number: 60966-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...