2'-Fluoro-2'-deoxyguanosine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64055-03
J64055-MD
AAJ64055-03EA
135
USD
AAJ64055-03
10814-310
2'-Fluoro-2'-deoxyguanosine 99%
2'-Fluoro-2'-deoxyguanosine
Powder
Purity: 99%
CAS No.: 78842-13-4
Molecular Formula: C10H12FN5O4
Formula Weight: 285.23
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00923832
Formula:
C₁₀H₁₂FN₅O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00923832 CAS Number: 78842-13-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...