Fluocinolone acetonide

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6alpha,9alpha-Difluoro-11beta,16alpha,17alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-Acetonide

J63996-03 J63996-MC
AAJ63996-03EA 288 USD
AAJ63996-03 AAJ63996-MC
Fluocinolone acetonide
Fluocinolone acetonide

A glucocorticoid with anti-inflammatory properties


Crystalline powder


Applications: A glucocorticoid with anti-inflammatory properties

Formula: C₂₄H₃₀F₂O₆
MW: 452.49 g/mol
Melting Pt: 267…269 °C
Storage Temperature: Ambient
MDL Number: MFCD00010525
CAS Number: 67-73-2
EINECS: 200-668-5
Merck Index: 14,4150

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR