Chicken Trypsin inhibitor (from Egg)

Supplier: Thermo Scientific Chemicals

J63927-03 J63927-04 J63927-ME
AAJ63927-03EA 218 USD
AAJ63927-03 AAJ63927-04 AAJ63927-ME
Chicken Trypsin inhibitor (from Egg)
Proteins and Peptides

Powder


CAS No.: 9035-81-8
Physical Form: Powder
Appearance: White
MDL No.: MFCD00082102


Applications: Inhibits the pancreatic enzyme trypsin

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR