Acebutolol hydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: N-(3-Acetyl-4-[2-hydroxy-3-(isopropylamino)propoxy]phenyl)butanamide, Acebutolol HCl, Acebutolol hydrochloride

J63790-09 J63790-03 J63790-06
AAJ63790-09EA 256 USD
AAJ63790-09 AAJ63790-03 AAJ63790-06
Acebutolol hydrochloride
Acebutolol hydrochloride

Cardioselective beta-adrenergic blocker


White to off-white powder


Applications: Cardioselective beta-adrenergic blocker

Formula: C₁₈H₂₈N₂O₄·HCl
MW: 372.89 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078860
CAS Number: 34381-68-5
EINECS: 251-980-3
Merck Index: 14,19

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR