Sotalol hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

J63772-03 J63772-06 J63772-MC
AAJ63772-03EA 520 USD
AAJ63772-03 AAJ63772-06 AAJ63772-MC
Sotalol hydrochloride 98%
Sotalol hydrochloride

Potent beta adrenergic antagonist


Powder


Applications: Potent beta adrenergic antagonist

Formula: C₁₂H₂₀N₂O₃S·HCl
Storage Temperature: Ambient
MDL Number: MFCD00242937
CAS Number: 959-24-0
EINECS: 213-496-0
Merck Index: 14,8728

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR