Imipramine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(10,11-Dihydro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine hydrochloride

J63723-06 J63723-14
AAJ63723-06EA 101.12 USD
AAJ63723-06 AAJ63723-14
Imipramine hydrochloride
Imipramine hydrochloride

A serotonin uptake inhibitor


Crystalline powder


Applications: A serotonin uptake inhibitor

Formula: C₁₉H₂₄N₂·HCl
MW: 316.87 g/mol
Melting Pt: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00012669
CAS Number: 113-52-0
EINECS: 204-030-7
Merck Index: 14,4920

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR