Ambroxol hydrochloride 98%

Supplier: Thermo Scientific Chemicals

J63712-06 J63712-03
AAJ63712-06EA 138 USD
AAJ63712-06 10814-296
Ambroxol hydrochloride 98%
Ambroxol hydrochloride

Sodium channel blocker. Metabolite of bromhexine


Powder


Applications: Sodium channel blocker. Metabolite of bromhexine

Formula: C₁₃H₁₈Br₂N₂O.HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00078932
CAS Number: 23828-92-4
EINECS: 242-899-2
Merck Index: 14,386

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR