DL-Cystine
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(H-DL-Cys-OH)2
J63564-06
J63564-14
J63564-22
AAJ63564-06EA
60.9
USD
AAJ63564-06
AAJ63564-14
AAJ63564-22
DL-Cystine
DL-Cystine
Powder
CAS No.: 923-32-0
Molecular Formula: C6H14N2O4S2
Formula Weight: 240.30
Physical Form: Powder
Appearance: White
Melting Point: 227° dec.
Beilstein Reference: 1728095
Merck Reference: 14,2782
MDL No.: MFCD00084652
Formula:
C₆H₁₂N₂O₄S₂ MW: 240.3 g/mol Boiling Pt: 429 °C (1013 hPa) Melting Pt: 227 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00084652 CAS Number: 923-32-0 EINECS: 213-094-5 Merck Index: 13,02811 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...