Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Uridine-5'-triphosphoric acid trisodium salt, Uridine triphosphate trisodium salt
J63427-MC
J63427-MD
AAJ63427-MCEA
107
USD
AAJ63427-MC
AAJ63427-MD
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3) ≥98%
Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3)
Powder
Purity: 98+%
CAS No.: 19817-92-6
Molecular Formula: C9H12N2Na3O15P3
Formula Weight: 550.09
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 3585234
MDL No.: MFCD00044310
Formula:
C₉H₁₂N₂Na₃O₁₅P₃ MW: 550.09 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00044310 CAS Number: 19817-92-6 EINECS: 243-347-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...