Gellan gum
Supplier: Thermo Scientific Chemicals
J63423-30
J63423-A1
AAJ63423-30EA
188.1
USD
AAJ63423-30
AAJ63423-A1
Gellan gum
Gellan gum
An agar substitute and gelling agent
Powder
An agar substitute and gelling agent
Storage Temperature:
Ambient |
MDL Number:
MFCD00131909 CAS Number: 71010-52-1 EINECS: 275-117-5 Merck Index: 14,4383 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...