Zeatin riboside

Supplier: Thermo Scientific Chemicals

Synonyms: trans-Zeatinriboside, trans-Zeatin-riboside

J63382-MB J63382-MC J63382-MF
AAJ63382-MBEA 198 USD
AAJ63382-MB AAJ63382-MC AAJ63382-MF
Zeatin riboside
Zeatin riboside

Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence


Powder


Applications: Main effects in tissue culture systems include control of shoot growth and formation, cell division and inhibition of leaf senescence

Formula: C₁₅H₂₁N₅O₅
MW: 351.363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036809
CAS Number: 6025-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR