1,1-Dimethylbiguanide hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

J63361-06 J63361-14 J63361-22
AAJ63361-06EA 45.01 USD
AAJ63361-06 AAJ63361-14 AAJ63361-22
1,1-Dimethylbiguanide hydrochloride
1,1-Dimethylbiguanide hydrochloride

An antidiabetic agent


Powder


An antidiabetic agent

Formula: C₄H₁₂ClN₅
Melting Pt: 218…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00012582
CAS Number: 1115-70-4
EINECS: 214-230-6
Merck Index: 14,5938

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR