3,3',5'-Triiodo-L-thyronine sodium salt
Supplier: Thermo Scientific Chemicals
Synonyms:
Liothyronine sodium salt, Sodium (S)-2-amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoate
J63312-03
J63312-ME
AAJ63312-03EA
249
USD
AAJ63312-03
AAJ63312-ME
3,3',5'-Triiodo-L-thyronine sodium salt
3,3',5'-Triiodo-L-thyronine sodium salt
Powder
CAS No.: 55-06-1
Molecular Formula: C15H11I3NNaO4
Formula Weight: 672.96
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White to pale brown
Melting Point: 205°
Beilstein Reference: 8179867
MDL No.: MFCD00002594
Formula:
C₁₅H₁₁I₃NNaO₄ MW: 672.959 g/mol Melting Pt: 205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002594 CAS Number: 55-06-1 EINECS: 200-223-5 Merck Index: 14,5510 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...