Cephalexin, hydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(6R,7R)-7-((R)-2-amino-2-phenylacetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(D-α-amino-phenylacetamido)-3-methyl-3-cepheme-4-carboxylic acid
J63172-06
J63172-14
AAJ63172-06EA
82.86
USD
AAJ63172-06
AAJ63172-14
Cephalexin, hydrate 97+%
Cephalexin
Colorimetric substrate for chymotrypsin, human leukocyte cathepsin G, and peptidyl prolyl isomerase
Powder
Applications: Colorimetric substrate for chymotrypsin, human leukocyte cathepsin G, and peptidyl prolyl isomerase
Formula:
C₁₆H₁₉N₃O₅S MW: 365.404 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00167148 CAS Number: 15686-71-2 EINECS: 239-773-6 Merck Index: 14,1974 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...