PIPES-Buffered Saline (5X), pH 6.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63152-AK
J63152-AP
AAJ63152-AKEA
56.8
USD
AAJ63152-AK
AAJ63152-AP
PIPES-Buffered Saline (5X), pH 6.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 5625-37-6
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00006159
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...