Thapsigargin 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-(((Z)-2-methylbut-2-enoyl)oxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate

J62866-MA J62866-LB0 J62866-MCR
AAJ62866-MAEA 820 USD
AAJ62866-MA AAJ62866-LB0 AAJ62866-MCR
Thapsigargin 95%
Thapsigargin

Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase


Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase


Cell permeable sesquiterpene lactone tumor promoter. Inhibits endoplasmic reticulum calcium-ATPase

Formula: C₃₄H₅₀O₁₂
MW: 650.762 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083511
CAS Number: 67526-95-8
Merck Index: 14,9272

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR