TRIS-buffered saline (TBS, 10X, high salt) pH 7.4, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62664-AP J62664-K2 J62664-K3
AAJ62664-APEA 62.13 USD
AAJ62664-AP AAJ62664-K2 AAJ62664-K3
TRIS-buffered saline (TBS, 10X, high salt) pH 7.4, Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR