Tylosin tartrate 94%

Supplier: Thermo Scientific Chemicals
Danger

J62633-03 J62633-06 J62633-09
AAJ62633-03EA 63.6 USD
AAJ62633-03 AAJ62633-06 AAJ62633-09
Tylosin tartrate 94%
Tylosin tartrate

Macrolide antibiotic


Powder


Applications: Macrolide antibiotic

Formula: C₄₆H₇₇NO₁₇·C₄H₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00070094
CAS Number: 1405-54-5
EINECS: 215-781-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR