Carbamazepine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5H-Dibenzo[b,f]azepine-5-carboxamide

J62590-14 J62590-22
AAJ62590-14EA 132.79 USD
AAJ62590-14 AAJ62590-22
Carbamazepine 98%
Carbamazepine

Cytochrome P450 3A4 inducing anti-epileptic drug; ligand for the GABA-A receptor benzodiazepine modulatory site


Powder


Cytochrome P450 3A4 inducing anti-epileptic drug; ligand for the GABA-A receptor benzodiazepine modulatory site

Formula: C₁₅H₁₂N₂O
MW: 236.27 g/mol
Melting Pt: 189…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00005073
CAS Number: 298-46-4
EINECS: 206-062-7
Merck Index: 14,1781

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR