Fast Blue BB base

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4'-Amino-2',5'-diethoxybenzanilide, N-(4-Amino-2,5-diethoxyphenyl)benzamide, Azoic Diazo No. 20, Fast blue BB, C.I. 37175, Azoic diazo component 20 (base)

J62456-14 J62456-22
AAJ62456-14EA 50.8 USD
AAJ62456-14 AAJ62456-22
Fast Blue BB base
Fast Blue BB base

C.I. 37175


Powder


Applications: C.I. 37175

Formula: C₁₇H₂₀N₂O₃
MW: 300.36 g/mol
Melting Pt: 95…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00009091
CAS Number: 120-00-3
EINECS: 204-363-8
UN: 3143
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR