TE Buffer, (20X), pH 8.0, Autoclaved, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J62388-AK J62388-AP
AAJ62388-AKEA 109.47 USD
AAJ62388-AK AAJ62388-AP
TE Buffer, (20X), pH 8.0, Autoclaved, Thermo Scientific
Buffers Biological Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00236359

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR