Imetit dihydrobromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J62346-MC
J62346-MF
AAJ62346-MCEA
655
USD
AAJ62346-MC
AAJ62346-MF
Imetit dihydrobromide 98%
Imetit dihydrobromide
An extremely potent, high affinity agonist at H3 and H4 receptors
Powder
Applications: An extremely potent, high affinity agonist at H3 and H4 receptors
Formula:
C₆H₁₀N₄S·2HBr MW: 332.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153816 CAS Number: 32385-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...