L-Cystine dihydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl

J62292-14 J62292-22 J62292-30
AAJ62292-14EA 71 USD
AAJ62292-14 AAJ62292-22 AAJ62292-30
L-Cystine dihydrochloride 99%
L-Cystine dihydrochloride

Powder


Purity: 99% CAS No.: 30925-07-6 Molecular Formula: C6H12N2O4S2.2HCl Formula Weight: 313.22 Physical Form: Powder Appearance: White to pale yellow Melting Point: 228-232° MDL No.: MFCD00070399

Formula: C₆H₁₂N₂O₄S₂·2HCl
MW: 313.23 g/mol
Melting Pt: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00070399
CAS Number: 30925-07-6
EINECS: 250-391-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR