Erythromycin cell culture reagent
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(-)-Erythromycin
J62279-09
J62279-14
AAJ62279-09EA
191.56
USD
AAJ62279-09
AAJ62279-14
Erythromycin cell culture reagent
Erythromycin
Macrolide antibiotic that inhibits bacterial protein synthesis.
Crystalline powder
Macrolide antibiotic that inhibits bacterial protein synthesis
Formula:
C₃₇H₆₇NO₁₃ MW: 733.95 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00084654 CAS Number: 114-07-8 EINECS: 204-040-1 Merck Index: 14,3681 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...