Tamsulosin hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

J61999-MA J61999-MB J61999-MC
AAJ61999-MAEA 126 USD
AAJ61999-MA AAJ61999-MB AAJ61999-MC
Tamsulosin hydrochloride ≥98%
Tamsulosin hydrochloride

An alpha-1 adrenoceptor antagonist


Powder


Applications: An alpha-1 adrenoceptor antagonist

Formula: C₂₀H₂₈N₂O₅S
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00922997
CAS Number: 106463-17-6
Merck Index: 14,9049

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR