Sodium alginate, high viscosity

Supplier: Thermo Scientific Chemicals

Synonyms: Alginic acid sodium salt, Algin

J61887-22 J61887-30 J61887-A1
AAJ61887-22EA 133.59 USD
AAJ61887-22 AAJ61887-30 AAJ61887-A1
Sodium alginate, high viscosity
Sodium alginate

A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies


Powder


A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3
Merck Index: 14,241

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR