Dextromethorphan hydrobromide ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J61732-06
J61732-09
J61732-18
AAJ61732-06EA
115
USD
AAJ61732-06
AAJ61732-09
AAJ61732-18
Dextromethorphan hydrobromide ≥98%
Dextromethorphan hydrobromide
Solid
CAS No.: 125-69-9
Molecular Formula: C18H25NO.HBr
Formula Weight: 352.32
Physical Form: Solid
Appearance: White
Melting Point: 125-127°
Beilstein Reference: 6453793
MDL No.: MFCD02173901
Formula:
C₁₈H₂₆BrNO Melting Pt: 125…127 °C Storage Temperature: Ambient |
CAS Number:
125-69-9 EINECS: 204-750-1 Merck Index: 14,8091 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...